missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Molindone.HCl, Lipomed
Description
Molindone.HCl
Specifications
Specifications
| Kemisk navn eller materiale | Molindone Hydrochloride |
| CAS | 15622-65-8 |
| Analyt- eller komponentnavne | Molindone Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C16 H24 N2 O2 . Cl H |
| Synonym | 4 H-Indol-4-one, 3-ethyl-1,5,6,7-tetrahydro-2-methyl-5-(4-morpholinylmethyl)-, hydrochloride (1:1), 4 H-Indol-4-one, 3-ethyl-1,5,6,7-tetrahydro-2-methyl-5-(4-morpholinylmethyl)-, monohydrochloride (9 CI), Indol-4(5 H)-one, 3-ethyl-6,7-dihydro-2-methyl-5-(morpholinomethyl)-, monohydrochloride (8 CI), (±)-Molindone hydrochloride, EN 1733 A, Lindone, Moban, Molindone hydrochloride |
| InChI formel | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMIL | Cl.CCc1c(C)[nH]c2CCC(CN3CCOCC3)C(=O)c12 |
| IUPAC navn | 3-ethyl-2-methyl-5-(morpholin-4-ylmethyl)-1,5,6,7-tetrahydroindol-4-one;hydrochloride |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?