Få mere at vide
Molecular sieves, tantalum oxide based; mesoporous, 22 Angstrom, powder, Thermo Scientific™
Mængde:
250 mg
1 g
5 g
Pakningsstørrelse:
1g
250mg
5g
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Molecular sieves, tantalum oxide based |
| Navn note | Mesoporous |
| Partikelstørrelse | 22Å |
| Følsomhed | Hygroscopic |
| Mængde | 250 mg |
| Fysisk form | Powder |
| MDL nummer | MFCD00131613 |
| Synonym | Ta TMSI |