missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Sieves, 13X, Honeywell Fluka™
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Mesh Size | 4 to 8mesh |
Packaging | Poly Bottle |
Quantity | 1 kg |
Physical Form | Beads |
Linear Formula | Na86[AlO2)86(SiO2)106] · xH2O |
UN Number | NONH for all modes of transport |
Chemical Name or Material | Molecular Sieves, 13X |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'