Få mere at vide
Molecular Sieve Dehydrate with Indicator For Drying Gases, Beads, Honeywell Fluka™
Shop alle Honeywell Fluka produkter
Mængde:
250 g
1 kg
Pakningsstørrelse:
1kg
250g
Beskrivelse
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Molecular Sieve Dehydrate Fluka With Indicator For Drying Gases |
| Tæthed | 2g/cm |
| Emballage | Glass Bottle |
| Porestørrelse | 4 |
| Mængde | 250 g |
| CAS | 1318-02-1, 10124-43-3 |
| FN nummer | NONH for all modes of transport |