Learn More
Invitrogen™ SphingoStrips™ Membranes
For protein identification
Brand: Invitrogen™ S23753
6718.40 DKK valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Additional Details : Weight : 0.00450kg
Description
SpingoStrips™ membranes are designed for the identification of proteins possessing phosphoinositide or sphingolipid recognition domains and for analysis of their lipid-binding specificities.
Cell Analysis, Cell Structure, Enzyme and Protein Activity Assays, Lipase and Phopholipase Activity, Membranes (General) and Lipids, Plasma Membrane, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room temperature
Specifications
Store in refrigerator 2°C to 8°C and protect from light. | |
Membrane | |
Room Temperature |
Lipid Arrays | |
SphingoStrips™ |
For Research Use Only. Not for use in diagnostic procedures.