Få mere at vide
Invitrogen™ Pro-Q™ Diamond Phosphoprotein Enrichment Kit
Beskrivelse
The Pro-Q™ Diamond Phosphoprotein Enrichment Kit enables efficient, nonradioactive isolation of phosphoproteins from complex cellular extracts. The kit provides resin, reagents, and columns designed to isolate phosphoproteins from 0.5-1.0 mg of total cellular protein per column.
Protein and Peptide Purification, Protein Enrichment, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis
Tekniske data
Tekniske data
| Produkttype | Phosphoprotein Enrichment Kit |
| Produktlinje | PRO-Q |
| Mængde | 1 kit |