Learn More
Invitrogen™ Lipopolysaccharides From Escherichia coli Serotype 055:B5, Alexa Fluor™ 594 Conjugate
Bright, red-fluorescent conjugate can be used to follow lipopolysaccharide binding, transport and cell-internalization processes
Brand: Invitrogen™ L23353
1866.76 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.00525kg
Description
Cell Analysis, Cell Cycle, Cell Structure, Cell Viability, Proliferation and Function, Endocytosis, Exocytosis and Phagocytosis, Lysosomes, Vacuoles
Order Info
Shipping Condition: Room Temperature
Specifications
Store in freezer (-5 to -30°C) and protect from light. | |
Lipopolysaccharide Conjugate | |
Solid | |
Room Temperature |
Fluorescence | |
Alexa Fluor Dyes | |
Alexa Fluor™ | |
590/617 |
For Research Use Only. Not for use in diagnostic procedures.