Learn More
Invitrogen™ JC-9 Dye (Mitochondrial Membrane Potential Probe)
Mitochondrial membrane potential probe
Brand: Invitrogen™ D22421
6288.24 DKK valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Additional Details : Weight : 0.00525kg
Description
The ratiometric mitochondrial membrane potential probe JC-9 exhibits green fluorescence that is independent of membrane potential and an increase in red fluorescence at hyperpolarized membrane potentials.
Apoptosis, Cell Analysis, Cell Structure, Cell Viability & Cytotoxicity, Cell Viability, Proliferation & Function, Mitochondria, Mitochondria Function, Mitochondrial Function
Order Info
Shipping Condition: Room Temperature
Specifications
Green | |
Fluorescent | |
Mitochondrial Membrane Potential Probe | |
Mitochondria |
Store at room temperature and protect from light. | |
Fluorescence Microscope, Flow Cytometer | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.