Få mere at vide
Invitrogen™ Image-IT™ LIVE Intracellular Membrane and Nuclear Labeling Kit (Counterstains For GFP-Expressing Cells)
Beskrivelse
The Image-iT LIVE Intracellular Membrane and Nuclear Labeling Kit provides two stains - red-fluorescent CellTrace™ BODIPY™ TR methyl ester and blue-fluorescent Hoechst 33342 dye - for highly selective staining of the intracellular membranes and nuclei of live green-fluorescent protein (GFP)-transfected cells and tissues.
Cell Analysis, Cell Structure, Cell-Based Reporter Assays, Cellular Imaging, Enzyme & Protein Activity Assays, Fluorescent Protein (e.g. GFP) Assays, Fluorescent Protein Assays (GFP), Immunofluorescence (IF), Immunofluorescence Staining & Detection, Nucleus, Nucleoli & Nuclear Envelope, Plasma Membrane, Proteins, Expression, Isolation & Analysis, Reporter Assays
Ordreinfo
Shipping Condition: Room Temperature
Tekniske data
Tekniske data
| Farve | Red, Blue |
| Indhold og opbevaring | Store in freezer -5°C to -30°C and protect from light. |
| Til brug med (udstyr) | Fluorescence Microscope |
| Mængde | 1 sæt |
| Detektionsmetode | Fluorescence |
| Produktlinje | Image-iT |
| Forsendelsestilstand | Room Temperature |
| Produkttype | Membrane and Nuclear Labeling |
| Subcellulær lokalisering | Cell Membranes, Lipids, Nucleus |
For Research Use Only. Not for use in diagnostic procedures.
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.