Få mere at vide
Invitrogen™ Molecular Probes™ Handbook: A Guide to Fluorescent Probes and Labeling Technologies, 11th Edition
Artikelnummer. p-4926817
Format:
Hardcover
Softcover
Pakningsstørrelse:
Hver
Artikelnummer. 10420330
Brand: Invitrogen™ H37126
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
An essential comprehensive fluorescent labeling and detection reference for cell and molecular biology researchers
  • Use this extensively updated edition to optimize results for fluorescent labeling and detection experiments
  • Comprehensive updates with over 3000 technology solutions
  • Extensive scientific references—hundreds of new primary literature references to product applications
  • Reorganized content—fresh design for easier access to data, structures, and information
  • New technical notes and product highlights
  • Available in hardcover and softcover
Applications:
Cell Analysis, Cell Structure, Cell Tracing & Tracking, Cell Viability, Proliferation & Function, Cellular Imaging, Flow Cytometry, Labeling Chemistry
TRUSTED_SUSTAINABILITY
Tekniske data
Format Hardcover