Få mere at vide
Invitrogen™ Molecular Probes™ Handbook: A Guide to Fluorescent Probes and Labeling Technologies, 11th Edition
Artikelnummer. 10511110
Skift visning
Format:
Blødt omslag
Indbundet
Pakningsstørrelse:
Hver
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Format Pakningsstørrelse
10511110 Blødt omslag Hver
10420330 Indbundet Hver
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 10511110 Leverandør Invitrogen™ Leverandørnr. H37127
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
An essential comprehensive fluorescent labeling and detection reference for cell and molecular biology researchers
  • Use this extensively updated edition to optimize results for fluorescent labeling and detection experiments
  • Comprehensive updates with over 3000 technology solutions
  • Extensive scientific references—hundreds of new primary literature references to product applications
  • Reorganized content—fresh design for easier access to data, structures, and information
  • New technical notes and product highlights
  • Available in hardcover and softcover
Applications:
Cell Analysis, Cell Structure, Cell Tracing & Tracking, Cell Viability, Proliferation & Function, Cellular Imaging, Flow Cytometry, Labeling Chemistry
TRUSTED_SUSTAINABILITY
Tekniske data
Format Blødt omslag