Få mere at vide
Invitrogen™ Molecular Probes™ Handbook: A Guide to Fluorescent Probes and Labeling Technologies, 11th Edition
Description
- Use this extensively updated edition to optimize results for fluorescent labeling and detection experiments
- Comprehensive updates with over 3000 technology solutions
- Extensive scientific references—hundreds of new primary literature references to product applications
- Reorganized content—fresh design for easier access to data, structures, and information
- New technical notes and product highlights
- Available in hardcover and softcover
Applications:
Cell Analysis, Cell Structure, Cell Tracing & Tracking, Cell Viability, Proliferation & Function, Cellular Imaging, Flow Cytometry, Labeling Chemistry
Specifications
Specifications
| Format | Indbundet |