Få mere at vide
Invitrogen™ Molecular Probes™ Epidermal Growth Factor (EGF) (from Mouse Submaxillary Glands)
Artikelnummer. 11508956
Mængde:
100 μg
Pakningsstørrelse:
100µg
Artikelnummer. 11508956
Brand: Invitrogen™ E3476
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Epidermal growth factor (EGF) is a 53 amino acid (6,045 Da) protein
  • Unlabeled version may be used as competitor molecule (control) in experiments that use labeled EGF to study receptor binding
Cell Analysis, Cell Viability, Proliferation and Function, Cellular Imaging, Endocytosis, Exocytosis and Phagocytosis, Immunofluorescence (IF), Immunofluorescence Staining and Detection
Ordreinfo
Shipping Condition: Wet Ice
TRUSTED_SUSTAINABILITY
Tekniske data
Mængde 100 μg