Få mere at vide
Invitrogen™ Carboxy-PTIO (spin trap)
Artikelnummer. 11500176
Mængde:
25 mg
Pakningsstørrelse:
25mg
Artikelnummer. 11500176
Brand: Invitrogen™ C7912
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Stable free radical that reacts stoichiometrically with nitric oxide
  • Water-soluble carboxy-PTIO has been used as nitric oxide antagonist as well as to quantitate NO levels by ESR spectrometry
Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Nitric Oxide Research, Nitro-Oxidative Stress
Ordreinfo
Shipping Condition: Room Temperature