Få mere at vide
Invitrogen™ 5-Dodecanoylaminofluorescein
Mængde:
100 mg
Pakningsstørrelse:
100mg
Beskrivelse
The lipophilic fluorescein probe, 5-dodecanoylaminofluorescein contains a C12 alkyl chain that will bind to membranes with the fluorophore at the aqueous interface and the alkyl tail protruding into the lipid interior.
Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection
Ordreinfo
Shipping Condition: Room Temperature
Tekniske data
Tekniske data
| Indhold og opbevaring | Store at room temperature and protect from light. |
| Mængde | 100 mg |
| Reagenstype | Fatty Acids |
| Forsendelsestilstand | Room Temperature |
| Etikettype | Classic Dyes |
| Produkttype | Lipophilic Probe |
For Research Use Only. Not for use in diagnostic procedures.