Få mere at vide
Moenomycin Complex (~5-10% Over CaCO3), TRC
Artikelnummer. 30008416
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30008416 5 mg
30009157 10 mg
30006371 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30008416 Leverandør Toronto Research Chemicals Leverandørnr. TRCM4853835MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 11015-37-5
Molekylær formel C69 H107 N4 O35 P
Molekylvægt (g/mol) 1583.57
Synonym Bambermycin (9CI, ACI), Moenomycin (8CI), Flapol, Flavofosfolipol, Flavomycin, Flavophospholipol, Gainpro, Menomycin, MeSH ID: D001455, Pharmastim
IUPAC navn (2S,3S,4R,5R,6R)-5-[(2S,3R,4R,5S,6R)-3-acetamido-5-[(2S,3R,4R,5S)-3-acetamido-4-hydroxy-6-methyl-5-[(2R,3R,4S,5R,6S)-3,4,5-trihydroxy-6-[(2-hydroxy-5-oxocyclopenten-1-yl)carbamoyl]oxan-2-yl]oxyoxan-2-yl]oxy-4-hydroxy-6-[[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxymethyl]oxan-2-yl]oxy-4-carbamoyloxy-6-[[(2R)-2-carboxy-2-[(2E,6E,13E)-3,8,8,14,18-pentamethyl-11-methylidenenonadeca-2,6,13,17-tetraenoxy]ethoxy]-hydroxyphosphoryl]oxy-3-hydroxy-3-methyloxane-2-carboxylic acid
SMIL CC1O[C@@H](O[C@H]2[C@H](O)[C@@H](NC(=O)C)[C@H](O[C@H]3[C@@H](OP(=O)(O)OC[C@@H](OC\C=C(/C)\CC\C=C\C(C)(C)CCC(=C)C\C=C(/C)\CCC=C(C)C)C(=O)O)O[C@H](C(=O)O)[C@@](C)(O)[C@@H]3OC(=O)N)O[C@@H]2CO[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)[C@H](NC(=O)C)[C@@H](O)[C@@H]1O[C@@H]5O[C@@H]([C@H](O)[C@H](O)[C@H]5O)C(=O)NC6=C(O)CCC6=O
Tekniske data
Kemisk navn eller materiale Flavomycin
Mængde 5 mg
InChI formel InChI=1S/C69H107N4O35P/c1-30(2)15-14-17-31(3)18-19-33(5)22-25-68(9,10)24-13-12-16-32(4)23-26-96-41(60(88)89)29-98-109(94,95)108-66-56(57(107-67(70)92)69(11,93)58(106-66)61(90)91)105-63-44(72-36(8)76)47(81)54(40(101-63)28-97-64-51(85)48(82)45(79)39(27-74)100-64)103-62-43(71-35(7)75)46(80)53(34(6)99-62)102-65-52(86)49(83)50(84)55(104-65)59(87)73-42-37(77)20-21-38(42)78/h13,15,18,23-24,34,39-41,43-58,62-66,74,77,79-86,93H,5,12,14,16-17,19-22,25-29H2,1-4,6-11H3,(H2,70,92)(H,71,75)(H,72,76)(H,73,87)(H,88,89)(H,90,91)(H,94,95)/b24-13+,31-18+,32-23+/t34?,39-,40-,41-,43-,44-,45-,46-,47-,48+,49+,50-,51-,52-,53-,54-,55+,56-,57-,58-,62+,63+,64-,65-,66-,69+/m1/s1
Formel vægt 1582.6453