Learn More
Ansell Model 111- The AlphaTec™ 1500 PLUS Stitched Coverall
Product Code. 30259534 Shop All Ansell Products
Change view
Click to view available options
Size:
8
9
Unit Size:
Pack of 30
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
30259534 9 Pack of 30
30259519 8 Pack of 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30259534 Supplier Ansell Supplier No. BL15S0011109
Forventet forsendelse: 04-05-2027
Particle-filtering, anti-static SMS protective suit, delivering light chemical resistance
  • Specialized protection: The AlphaTec™ 1500 PLUS Stitched - Model 111 delivers light chemical resistance, guards against asbestos and filters 99.9% of particles over 3 microns in size, as well as brick and cement dust, light aerosols and mists
  • Heightened breathability: This asbestos protection suit's air and water vapor permeability also reduces wearers' exposure to heat stress
  • Enhanced fit and features: Wearer comfort is augmented by this protective body suit's optimal fit, while a three-piece hood offers additional protective coverage, finger loops offer a secure hold.
  • Certified antistatic capabilities: The AlphaTec™ 1500 PLUS Stitched - Model 111 is tested against EN 1149-5 antistatic standards
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hooded Coveralls
Size 9
Gender Unisex
Material Anti-static SMS
Color Blue
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Finger Loops, Elastic
Neck Style Hooded
Hood Type 3-panel, Elastic
Closure Type 2-way Front Zipper with Storm Flap and Finger Loops
Seam Type Stitched
Sterility Non-sterile
Resistance Chemicals, Particle, Anti-static