Få mere at vide
Modafinil-d10 Carboxylate, TRC
Artikelnummer. 30482306
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30482306 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30482306 Leverandør Toronto Research Chemicals Leverandørnr. TRCM48251310MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Modafinil-d10 Carboxylate
CAS 1246819-82-8
Molekylær formel C15H4D10O3S
Mængde 10 mg
Synonym 2-[(Diphenyl-methyl)sulfinyl-d10]acetic Acid, Modafinil Acid-d10, Modafinic Acid-d10
InChI formel InChI=1S/C15H14O3S/c16-14(17)11-19(18)15(12-7-3-1-4-8-12)13-9-5-2-6-10-13/h1-10,15H,11H2,(H,16,17)/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D
SMIL [2H]C1=C(C(=C(C(=C1[2H])[2H])C(C2=C(C(=C(C(=C2[2H])[2H])[2H])[2H])[2H])S(=O)CC(=O)O)[2H])[2H]
IUPAC navn 2-[bis(2,3,4,5,6-pentadeuteriophenyl)methylsulfinyl]acetic acid
Molekylvægt (g/mol) 284.4
Formel vægt 284.1291