Få mere at vide
Modafinil Carboxylate Methyl Ester, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methyl [(RS)-(Diphenylmethyl)sulfinyl]acetate |
| CAS | 63547-25-1 |
| Molekylær formel | C16 H16 O3 S |
| Mængde | 25 mg |
| Synonym | Methyl [(RS)-(Diphenylmethyl)sulfinyl]acetate, Modafinil Imp. C (EP) |
| InChI formel | InChI=1S/C16H16O3S/c1-19-15(17)12-20(18)16(13-8-4-2-5-9-13)14-10-6-3-7-11-14/h2-11,16H,12H2,1H3 |
| SMIL | COC(=O)CS(=O)C(c1ccccc1)c2ccccc2 |
| IUPAC navn | methyl 2-benzhydrylsulfinylacetate |
| Molekylvægt (g/mol) | 288.36 |
| Formel vægt | 288.082 |