Få mere at vide
Mocetinostat, MedChemExpress
Artikelnummer. 16431247
Encompass_Preferred
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16431247 50 mg
16415008 10 mg
16415766 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16431247 Leverandør Medchem Express Leverandørnr. HY12164
MedChemExpress Mocetinostat (MGCD0103) is a potent, orally active and isotype-selective HDAC (Class I/IV) inhibitor with IC50s of 0.15, 0.29, 1.66 and 0.59 μM for HDAC1, HDAC2, HDAC3 and HDAC11, respectively. Mocetinostat shows no inhibition on HDAC4, HDAC5, HDAC6, HDAC7, or HDAC8.
Mocetinostat (MGCD0103) is a potent, orally active and isotype-selective HDAC (Class I/IV) inhibitor with IC50s of 0.15, 0.29, 1.66 and 0.59 μM for HDAC1, HDAC2, HDAC3 and HDAC11, respectively. Mocetinostat shows no inhibition on HDAC4, HDAC5, HDAC6, HDAC7, or HDAC8.
Tekniske data
Kemisk navn eller materiale Mocetinostat
Fysisk form Powder
CAS 726169-73-9
Farve Off-White
Mængde 50 mg
Renhedsgrad noter Research
Molekylær formel C23H20N6O
Til brug med (applikation) Cancer-programmed cell death
Synonym MGCD0103
Opløselighedsinformation DMSO : 50 mg/mL (126.12 mM; Need ultrasonic)
SMIL O=C(C1=CC=C(C=C1)CNC2=NC=CC(C3=CC=CN=C3)=N2)NC4=CC=CC=C4N
Molekylvægt (g/mol) 396.44
Formel vægt 396.44
Procent renhed 98.92%