Learn More
Miele™ Mobile Injector Unit
Brand: Miele™ 3809490
Additional Details : Weight : 50.00000kg
Specifications
Mobile Injector Unit | |
Narrow-Necked Glassware | |
1 Each |
E 350 | |
15 x E 351 injector 4,0 x 160mm, 15 x E 353 clip for injector 4,0 x 160mm, 18 x E 352 injector 6,0 x 220mm, 18 x E 354 clip for injector 6,0 x 220mm, 1 x jet for flushing powder dispenser |