missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
STEMCELL Technologies MLN4924
Beskrivelse
- MLN4924 inhibits cullin neddylation by directly targeting the NEDD8-activating enzyme that is essential for the activity of cullin-RING ligase (CRL) (Nawrocki et al.) The components of CRL are frequently up-regulated in human cancers (Zhao & Sun), and MLN4924 was reported to suppress the growth of human tumor xenografts in mice (Soucy et al.)
- In combination with other small molecules ('CRISPY' mix), MLN4924 has been reported to increase precise CRISPR-Cas9 genome editing in human pluripotent stem cells (Riesenberg & Maricic)
- By blocking NAE and proteasome, MLN4924 together with bortezomib inhibits AKT and mTOR and induces apoptosis in multiple myeloma cell lines (Gu et al.)
Tekniske data
Tekniske data
| CAS | 905579-51-3 |
| Synonym | Pevonedistat |
| Mængde | 10 mg |
| Molekylær formel | C21H25N5O4S |
| Renhed | ≥98% |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?