Få mere at vide
Invitrogen™ MLKL Recombinant Rabbit Monoclonal Antibody (24GB4620)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557484
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
MLKL Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The protein kinase domain is predicted to be catalytically inactive. Molecular function: protein binding. There are two isoforms.
Tekniske data
| MLKL | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8NB16, Q9D2Y4 | |
| Mlkl | |
| A synthesized peptide derived from human MLKL (aa 371-472). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB4620 | |
| Unconjugated | |
| Mlkl | |
| 9130019I15Rik; hMLKL; mixed lineage kinase domain like pseudokinase; mixed lineage kinase domain-like; mixed lineage kinase domain-like protein; Mlkl; rCG51567-like | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 197259, 74568 | |
| -20°C | |
| Liquid |