missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLK2 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-06104IR
Additional Details : Weight : 0.00970kg
Description
MLK2 Polyclonal specifically detects MLK2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MLK2 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
EC 2.7.11, MEKK10, mitogen-activated protein kinase kinase kinase 10, Mixed lineage kinase 2, MKN28 kinase, MLK2MKN28 derived nonreceptor_type serine/threonine kinase, MSTEC 2.7.11.25, Protein kinase MST | |
Produced in rabbits immunized with E. coli-derived Human MLK2 fragment. | |
0.1 mL | |
Apoptosis | |
4294 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |