missing translation for 'onlineSavingsMsg'
Learn More

VistaLab™ Technologies MLA™ Silver Brand Precision Pipetters

Made of precision engineered aluminum alloy and stainless steel with limited Lifetime Warranty (covering parts). Individually serialized with Calibration Certificate included.

Brand:  VistaLab™ Technologies 1110

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 11975772

  • 1910.00 DKK / Each
Estimated Shipment: 08-07-2024
to see stock.
Add to basket

Description

Description

  • “To deliver” capability ensures volume accuracy over a wide range of viscosities
  • Ideal for repetitive delivery of reagents when high precision and accuracy are required
  • Capacity engraved on bonnet

Specifications

Specifications

±2%
Silver
MLA Precision
Non-sterile
90 μL
±0.57%
Limited Lifetime
2 in.
Not Autoclavable
Aluminum Alloy, Stainless Steel
1
Precision Pipetter
Nozzle Insert, Calibration Key
1 Ea.
9 in.
1.5 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers