Få mere at vide
ML327, MedChemExpress
Artikelnummer. 16484836
Encompass_Preferred
Skift visning
Mængde:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Pakningsstørrelse:
10mg
50mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16484836 100 mg -
16466507 5 mg -
16460989 1 mg -
16432856 25 mg -
16423788 10 mg 10mg
16420167 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Artikelnummer. 16484836 Leverandør Medchem Express Leverandørnr. HY103038
MedChemExpress ML327 is a blocker of MYC which can also de-repress E-cadherin transcription and reverse Epithelial-to-Mesenchymal Transition (EMT).
ML327 is a blocker of MYC which can also de-repress E-cadherin transcription and reverse Epithelial-to-Mesenchymal Transition (EMT).
Tekniske data
Kemisk navn eller materiale ML327
Fysisk form Solid
CAS 1883510-31-3
Farve White
Mængde 100 mg
Renhedsgrad noter Research
Molekylær formel C19H18N4O4
Til brug med (applikation) Cancer-programmed cell death
Opløselighedsinformation DMSO : 32 mg/mL (87.34 mM; Need ultrasonic and warming)
SMIL O=C(C1=CC=CNC1=O)NCCCNC(C2=NOC(C3=CC=CC=C3)=C2)=O
Molekylvægt (g/mol) 366.37
Formel vægt 366.37
Procent renhed 97.56%