missing translation for 'onlineSavingsMsg'
Få mere at vide

ML218, MedChemExpress

Artikelnummer. 16476547
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16476547 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16476547 Leverandør Medchem Express Leverandørnr. HY103309

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress ML218 is a potent, selective and orally active T-type Ca2+ channels (Cav3.1, Cav3.2, Cav3.3) inhibitor with IC50s of 310 nM and 270 nM for Cav3.2 and Cav3.3, respectively. ML218 inhibits the burst activity in subthalamic nucleus (STN) neurons. ML218 has no significant inhibition of L- or N-type calcium channels, KATP or hERG potassium channels. ML218 can penetrate the blood-brain barrier.

ML218 is a potent, selective and orally active T-type Ca2+ channels (Cav3.1, Cav3.2, Cav3.3) inhibitor with IC50s of 310 nM and 270 nM for Cav3.2 and Cav3.3, respectively. ML218 inhibits the burst activity in subthalamic nucleus (STN) neurons. ML218 has no significant inhibition of L- or N-type calcium channels, KATP or hERG potassium channels. ML218 can penetrate the blood-brain barrier.

Tekniske data

Kemisk navn eller materiale ML218
Fysisk form Solid
CAS 1346233-68-8
Farve Light Yellow
Mængde 5 mg
Renhedsgrad noter Research
Molekylær formel C19H26Cl2N2O
Til brug med (applikation) Neuroscience-Neuromodulation
Opløselighedsinformation DMSO : 125 mg/mL (338.45 mM; Need ultrasonic)
SMIL O=C(NC[C@@H]1[C@@]2([H])CN(CCC(C)(C)C)C[C@@]12[H])C3=CC(Cl)=CC(Cl)=C3
Molekylvægt (g/mol) 369.33
Formel vægt 369.33
Procent renhed 99.18%
Grad Research
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.