Få mere at vide
Tocris™ ML 402
Beskrivelse
ML 402 is a selective TREK channel activator. Selectively activates K2P2.1 (TREK-1) and K2P10.1 (TREK-2) with no significant effect at K2P4.1 channels (TRAAK).
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-[2-(4-Chloro-2-methylphenoxy)ethyl]-2-thiophenecarboxamide |
| CAS | 298684-44-3 |
| Mængde | 10 mg |
| Molekylær formel | C14H14ClNO2S |
| Renhed | 98% |