Få mere at vide
ML 311, Tocris Bioscience™
Inhibits Mcl-1-BIM interaction
Brand: Tocris Bioscience 5733/10
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
ML 311 is an inhibits Mcl1-BIM interaction. Inhibits viability of Mcl1-dependent tumor cell lines. Induces cell death and decreased spheroid formation in Her2 positive breast cancer cells.Tekniske data
| 7-[(4-Ethyl-1-piperazinyl)[4-(trifluoromethyl)phenyl]methyl]-8-quinolinol | |
| 315698-17-0 | |
| >99% | |
| 10 mg |