Få mere at vide
Tocris Bioscience™ ML 233, Tocris Bioscience™
Non-peptide apelin receptor agonist
Brand: Tocris Bioscience™ 5966/10
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
ML 233 is a non-peptide apelin receptor agonist (EC50 = 3.7 μM); exhibits >21-fold selectivity over the closely related angiotensin 1 receptor (EC50 = >79 μM).Tekniske data
| (E)-4-Methyl-5-(((phenylsulfonyl)oxy)imino)-[1,1'-bi(cyclohexane)]-3,6-dien-2-one | |
| 10 mg | |
| Solid | |
| Bioactive Small Molecule |
| 143621-35-6 | |
| Soluble to 50mM in DMSO | |
| White |
Produkttitel
Ser du en mulighed for forbedring?
For Research Use Only.