Få mere at vide
Tocris Bioscience™ ML 179
GREENER_CHOICE
Selective liver receptor homolog 1 (LRH1) inverse agonist
Brand:  Tocris Bioscience™ 4957/10
Encompass
Produktkode 15716605
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
ML 179 is a selective liver receptor homolog 1 (LRH1, NR5A2) inverse agonist (IC50 = 320 nM and maximum efficacy of 64% repression). Exhibits >31-fold selectivity over SF1.
Tekniske data
Tekniske data
ML 179
3-Cyclohexyl-6-[4-[3-(trifluoromethyl)phenyl]piperazin-1-yl]-1H-pyrimidine-2,4-dione
Soluble to 50mM in DMSO
Bioactive Small Molecule
1883548-87-5
10 mg
Crystalline Solid
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner
Produkttitel
Vælg et problem
Research Use Only. Not For Human or Veterinary Use