Få mere at vide
Tocris Bioscience™ MKT 077
GREENER_CHOICE
Artikelnummer. 30001679
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30001679 50 mg
18074012 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30001679 Leverandør Tocris Bioscience™ Leverandørnr. 4621/50
Binds mot-2; selectively cytotoxic in cancer cells
Occupies mortalin-2 (mot-2), a member of the Hsp70 family, at its p53 binding site and enables p53 translocation to the nucleus. Selectively cytotoxic; causes growth arrest of cancer cells in culture. Also inhibits telomerase activity and cross-links F-actin.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 147366-41-4
Molekylær formel C21H22ClN3OS2
Tekniske data
Kemisk navn eller materiale MKT 077
Mængde 50 mg
Formel vægt 432
Renhed >98%
Molekylvægt (g/mol) 432.00