Learn More
Invitrogen™ MKNK2 (MNK2) Human Protein
MKNK1 and MKNK2 are the Map kinase-interacting kinases.
Brand:  Invitrogen™ PR8046A
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15478643
  • 13290.00 DKK / 100µg
Estimated Shipment: 13-05-2024
to see stock.
Description
Description
The Map kinase-interacting kinases are activated by Map kinases, regulate protein synthesis by phosphroylation eIF4E on Ser209, and may contribute to tumorigenesis.
Specifications
Specifications
NP_060042.2
50mM Tris pH 7.5, 150mM NaCl, 0.02% Trition X-100, 0.5mM EDTA, 2mM DTT, 50% Glycerol.
100μg
RUO - research use only
MKNK2 (MNK2)
Human
Baculovirus
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
74.2kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
MAPKAPK Kinase Family
GST Tag
Frozen