missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MKNK1 (MNK1) Human Protein

MKNK1 and MKNK2 are the Map kinase-interacting kinases.

13300.00 DKK - 57500.00 DKK

Specifications

Accession Number NP_945324
For Use With (Application) Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
Molecular Weight (g/mol) 66.6kDa
Storage Requirements Kinase proteins are shipped on dry ice and should be stored at -80°C.
Regulatory Status RUO - research use only
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15488713
View Documents Promotion Details
Invitrogen™
PR9138A
100μg
13300.00 DKK
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 06-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15448803
View Documents Promotion Details
Invitrogen™
PV6024
1mg
57500.00 DKK
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 06-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Map kinase-interacting kinases are activated by Map kinases, regulate protein synthesis by phosphroylating eIF4E on Ser209, and may contribute to tumorigenesis.
Specifications

Specifications

NP_945324
66.6kDa
RUO - research use only
MKNK1 (MNK1)
Human
Insect
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
Kinase proteins are shipped on dry ice and should be stored at -80°C.
Full Length
MAPKAPK Kinase Family
GST Tag
Frozen
Videos
SDS
Documents

Documents

Product Certifications