Få mere at vide
Tocris Bioscience™ MK6-83
TRPML channel activator
Brand: Tocris Bioscience™ 5547/10
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
MK6-83 is a TRPML channel activator (EC50 = 110 nM). Also activates F408Δ and F465L TRPML mutants (EC50 values are 0.1 and 1.23 μM respectively). Restores endolysosomal trafficking and zinc homeostasis in lysosomes of mucolipidosis type IV (MLIV) mutant fibroblasts.Tekniske data
| MK6-83 | |
| 5-Methyl-N-[2-(1-piperidinyl)phenyl]-2-thiophenesulfonamide | |
| Soluble to 100mM in DMSO and to 20mM in ethanol | |
| Bioactive Small Molecule |
| 1062271-24-2 | |
| 10 mg | |
| Solid |
Produkttitel
Ser du en mulighed for forbedring?
Research Use Only. Not For Human or Veterinary Use