Få mere at vide
MK-571, TRC
Artikelnummer. 30474594
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30474594 10 mg
30465850 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30474594 Leverandør Toronto Research Chemicals Leverandørnr. TRCM42450010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 115104-28-4
Molekylær formel C26 H26 Cl N2 O3 S2 . Na
Molekylvægt (g/mol) 537.07
Synonym MK 571
IUPAC navn sodium;3-[[3-[(E)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl]-[3-(dimethylamino)-3-oxopropyl]sulfanylmethyl]sulfanylpropanoate
SMIL [Na+].CN(C)C(=O)CCSC(SCCC(=O)[O-])c1cccc(\C=C\c2ccc3ccc(Cl)cc3n2)c1
Tekniske data
Kemisk navn eller materiale MK-571
Mængde 10 mg
InChI formel InChI=1S/C26H27ClN2O3S2.Na/c1-29(2)24(30)12-14-33-26(34-15-13-25(31)32)20-5-3-4-18(16-20)6-10-22-11-8-19-7-9-21(27)17-23(19)28-22;/h3-11,16-17,26H,12-15H2,1-2H3,(H,31,32);/q;+1/p-1/b10-6+;
Formel vægt 536.0971