Få mere at vide
MK-0429, MedChemExpress
Artikelnummer. 16498897
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
Pakningsstørrelse:
100mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16498897 5 mg -
16446636 100 mg 100mg
16446318 10 mg -
16412437 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16498897 Leverandør Medchem Express Leverandørnr. HY15102
MedChemExpress MK-0429 (L-000845704) is an orally active, potent, selective and nonpeptide pan-integrin antagonist with IC50 values of 1.6 nM, 2.8 nM, 0.1 nM, 0.7 nM, 0.5 nM and 12.2 nM for αvβ1, αvβ3, αvβ5, αvβ6, αvβ8 and α5β1, respectively.
MK-0429 (L-000845704) is an orally active, potent, selective and nonpeptide pan-integrin antagonist with IC50 values of 1.6 nM, 2.8 nM, 0.1 nM, 0.7 nM, 0.5 nM and 12.2 nM for αvβ1, αvβ3, αvβ5, αvβ6, αvβ8 and α5β1, respectively.
Tekniske data
Kemisk navn eller materiale MK-0429
CAS 227963-15-7
Farve White
Renhedsgrad noter Research
Molekylær formel C23H29N5O4
Mængde 5 mg
Synonym L-000845704
Opløselighedsinformation DMSO : 100 mg/mL (227.53 mM; Need ultrasonic) ∣H2O : 2.78 mg/mL (6.33 mM; Need ultrasonic)
SMIL O=C1N([C@H](C2=CC=C(OC)N=C2)CC(O)=O)CCN1CCCC3=NC4=C(C=C3)CCCN4
Molekylvægt (g/mol) 439.51
Formel vægt 439.51
Procent renhed 98.33%
Grad Research
Fysisk form Powder
Til brug med (applikation) COVID-19-immunoregulation