Få mere at vide
Mitoxantrone-d8, TRC
Artikelnummer. 30464345
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30464345 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30464345 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3734271MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Mitoxantrone-d8
CAS 1189974-82-0
Molekylær formel C22 2H8 H20 N4 O6
Mængde 1 mg
Synonym Mitoxantrone D8
InChI formel InChI=1S/C22H28N4O6/c27-11-9-23-5-7-25-13-1-2-14(26-8-6-24-10-12-28)18-17(13)21(31)19-15(29)3-4-16(30)20(19)22(18)32/h1-4,23-30H,5-12H2/i9D2,10D2,11D2,12D2
SMIL [2H]C([2H])(O)C([2H])([2H])NCCNc1ccc(NCCNC([2H])([2H])C([2H])([2H])O)c2C(=O)c3c(O)ccc(O)c3C(=O)c12
IUPAC navn 1,4-dihydroxy-5,8-bis[2-[(1,1,2,2-tetradeuterio-2-hydroxyethyl)amino]ethylamino]anthracene-9,10-dione
Molekylvægt (g/mol) 452.53
Formel vægt 452.2511