Få mere at vide
Mitoquinol, TRC
Artikelnummer. 30469908
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30469908 50 mg
30483216 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30469908 Leverandør Toronto Research Chemicals Leverandørnr. TRCM37221050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 845959-55-9
Molekylær formel C38H49O7PS
Molekylvægt (g/mol) 680.83
Synonym [10-(3,6-Dihydroxy-4,5-dimethoxy-2-methylphenyl)decyl]triphenylphosphonium Methanesulfonate, 10-(2,5-Dihydroxy-3,4-dimethoxy-6-methylphenyl)decyl]triphenylphosphonium Methanesulfonate (1:1)
SMIL OC1=C(OC)C(OC)=C(O)C(C)=C1CCCCCCCCCC[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.CS(=O)([O-])=O
Tekniske data
Kemisk navn eller materiale Mitoquinol
Mængde 50 mg
InChI formel InChI=1S/C37H45O4P.CH4O3S/c1-29-33(35(39)37(41-3)36(40-2)34(29)38)27-19-8-6-4-5-7-9-20-28-42(30-21-13-10-14-22-30,31-23-15-11-16-24-31)32-25-17-12-18-26-32;1-5(2,3)4/h10-18,21-26H,4-9,19-20,27-28H2,1-3H3,(H-,38,39);1H3,(H,2,3,4)