Få mere at vide
Mirtazapine-d4, TRC
Artikelnummer. 30470424
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30470424 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30470424 Leverandør Toronto Research Chemicals Leverandørnr. TRCM36500310MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Mirtazapine-d4
CAS 1215898-55-7
Renhedsgrad noter HPLC
Molekylær formel C17H15D4N3
Mængde 10 mg
Synonym 1,2,3,4,10,14b-Hexahydro-2-methyl-pyrazino[2,1-a]pyrido[2,3-c][2]benzazepine-d4, Org-3770-d4, Remeron-d4, Zispin-d4
SMIL CN1C([2H])([2H])C([2H])([2H])N2C3=NC=CC=C3CC4=C(C=CC=C4)C2C1
Molekylvægt (g/mol) 269.38
Formel vægt 269.183
Procent renhed >95