Få mere at vide
Mirodenafil, LoGiCal
Artikelnummer. 30461095
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30461095 10 mg
1 options
Artikelnummer. 30461095 Leverandør LoGiCal Leverandørnr. LGCFOR3291.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Mirodenafil
CAS 862189-95-5
Analyt- eller komponentnavne Mirodenafil
Forsendelsestilstand Room Temperature
Molekylær formel C26 H37 N5 O5 S
Certificeringer/compliance ISO 9001
Synonym 4H-Pyrrolo[3,2-d]pyrimidin-4-one, 5-ethyl-3,5-dihydro-2-[5-[[4-(2-hydroxyethyl)-1-piperazinyl]sulfonyl]-2-propoxyphenyl]-7-propyl-, 1-Piperazineethanol, 4-[[3-(5-ethyl-4,5-dihydro-4-oxo-7-propyl-1H-pyrrolo[3,2-d]pyrimidin-2-yl)-4-propoxyphenyl]sulfonyl]- (9CI), Mirodenafil, Mvix
InChI formel InChI=1S/C26H37N5O5S/c1-4-7-19-18-30(6-3)24-23(19)27-25(28-26(24)33)21-17-20(8-9-22(21)36-16-5-2)37(34,35)31-12-10-29(11-13-31)14-15-32/h8-9,17-18,32H,4-7,10-16H2,1-3H3,(H,27,28,33)
Løsningstype Neat
SMIL CCCOc1ccc(cc1C2=Nc3c(CCC)cn(CC)c3C(=O)N2)S(=O)(=O)N4CCN(CCO)CC4
IUPAC navn 5-ethyl-2-[5-[4-(2-hydroxyethyl)piperazin-1-yl]sulfonyl-2-propoxyphenyl]-7-propyl-3H-pyrrolo[3,2-d]pyrimidin-4-one
Industry Type Clinical Testing
Molekylvægt (g/mol) 531.67
Mængde 10 mg
Formel vægt 531.2515 g/mol