missing translation for 'onlineSavingsMsg'
Få mere at vide

Minoxidil Sulfate, TRC

Artikelnummer. 30469738
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30469738 25 mg
30473071 5 mg
30470682 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30469738 Leverandør Toronto Research Chemicals Leverandørnr. TRCM34510025MG

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83701-22-8
Molekylær formel C9 H15 N5 O4 S
Molekylvægt (g/mol) 289.31
Synonym 2-Pyrimidinamine, 1,6-dihydro-6-imino-4-(1-piperidinyl)-1-(sulfooxy)-, Minoxidil sulfate
IUPAC navn (2,6-diamino-4-piperidin-1-ylpyrimidin-1-ium-1-yl) sulfate
SMIL Nc1cc(nc(N)[n+]1OS(=O)(=O)[O-])N2CCCCC2

Specifications

Kemisk navn eller materiale Minoxidil, Sulfate
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C9H15N5O4S/c10-7-6-8(13-4-2-1-3-5-13)12-9(11)14(7)18-19(15,16)17/h6H,1-5H2,(H4,10,11,12,15,16,17)
Formel vægt 289.0845
Procent renhed >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.