Få mere at vide
Minoxidil Sulfate-d10, TRC
Artikelnummer. 30481991
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30481991 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30481991 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3451025MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Minoxidil Sulfate-d10
CAS 1260373-92-9
Molekylær formel C9H5D10N5O4S
Mængde 5 mg
Synonym 6-(1-Piperidinyl-d10)-2,4-pyrimidinediamine-3-oxide Sulfate, 1,6-Dihydro-6-imino-4-(1-piperidinyl-d10)-1-(sulfooxy)-2-pyrimidinamine
InChI formel InChI=1S/C9H15N5O4S/c10-7-6-8(13-4-2-1-3-5-13)12-9(11)14(7)18-19(15,16)17/h6H,1-5H2,(H4,10,11,12,15,16,17)/i1D2,2D2,3D2,4D2,5D2
SMIL NC1=[N+](OS(=O)([O-])=O)C(N)=NC(N2C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C2([2H])[2H])=C1
Molekylvægt (g/mol) 299.37