missing translation for 'onlineSavingsMsg'
Få mere at vide

Minoxidil Sulfate-d10, TRC

Artikelnummer. 30471629
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30471629 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30471629 Leverandør Toronto Research Chemicals Leverandørnr. TRCM34510250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Minoxidil Sulfate-d10
CAS 1260373-92-9
Molekylær formel C9H5D10N5O4S
Mængde 50 mg
Synonym 6-(1-Piperidinyl-d10)-2,4-pyrimidinediamine-3-oxide Sulfate, 1,6-Dihydro-6-imino-4-(1-piperidinyl-d10)-1-(sulfooxy)-2-pyrimidinamine
InChI formel InChI=1S/C9H15N5O4S/c10-7-6-8(13-4-2-1-3-5-13)12-9(11)14(7)18-19(15,16)17/h6H,1-5H2,(H4,10,11,12,15,16,17)/i1D2,2D2,3D2,4D2,5D2
SMIL NC1=[N+](OS(=O)([O-])=O)C(N)=NC(N2C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C2([2H])[2H])=C1
Molekylvægt (g/mol) 299.37
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.