Learn More
Minoxidil, LoGiCal
Product Code. 30462439
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity Unit Size
30462439 10 mg 10mg
1 options
Product Code. 30462439 Supplier LoGiCal Supplier No. LGCFOR0521.00
 Restricted Item
Forventet forsendelse: 30-11-2026
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Minoxidil
CAS 38304-91-5
Analyte or Component Names Minoxidil
Shipping Condition Room Temperature
Molecular Formula C9 H15 N5 O
Certifications/Compliance ISO 9001
Synonym 2,4-Diamino-6-piperidinopyrimidine 3-N-oxide, 2,4-Diamino-6-piperidinopyrimidine 3-oxide, 6-Amino-1,2-dihydro-1-hydroxy-2-imino-4-piperidinopyrimidine, Alopexil, Alostil, Loniten, Lonolox, Minoxidil, Minoximen, Mintop, Normoxidil, PDP, Pierminox, Prexidil, RiUP, Rogaine, Tricoxidil, U 10858
InChI Formula InChI=1S/C9H15N5O/c10-7-6-8(12-9(11)14(7)15)13-4-2-1-3-5-13/h6H,1-5,10H2,(H2,11,12)
Solution Type Neat
SMILES Nc1cc(nc(N)[n+]1[O-])N2CCCCC2
IUPAC Name 3-oxido-6-piperidin-1-ylpyrimidin-3-ium-2,4-diamine
Industry Type Clinical Testing
Molecular Weight (g/mol) 209.25
Quantity 10 mg
Formula Weight 209.1277 g/mol