missing translation for 'onlineSavingsMsg'
Learn More

Minoxidil-d10, TRC

Product Code. 30482064
Change view
Click to view available options
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30482064 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30482064 Supplier Toronto Research Chemicals Supplier No. TRCM34500225MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Minoxidil-d10
CAS 1020718-66-4
Molekylær formel C9 2H10 H5 N5 O
Mængde 25 mg
Synonym 2,4-Pyrimidinediamine, 6-(1-piperidinyl-2,2,3,3,4,4,5,5,6,6-d10)-, 3-oxide (ACI), 6-(1-Piperidinyl-2,2,3,3,4,4,5,5,6,6-d10)-2,4-pyrimidinediamine 3-oxide, 6-(1-Piperidinyl-d10)-2,4-pyrimidinediamine 3-oxide, Minoxidil-d10, Minoxidil-D10 (piperidine-D10)
InChI formel InChI=1S/C9H15N5O/c10-7-6-8(12-9(11)14(7)15)13-4-2-1-3-5-13/h6H,1-5,10H2,(H2,11,12)/i1D2,2D2,3D2,4D2,5D2
SMIL [2H]C1([2H])N(c2cc(N)[n+]([O-])c(N)n2)C([2H])([2H])C([2H])([2H])C([2H])([2H])C1([2H])[2H]
IUPAC navn 6-(2,2,3,3,4,4,5,5,6,6-decadeuteriopiperidin-1-yl)-3-oxidopyrimidin-3-ium-2,4-diamine
Molekylvægt (g/mol) 219.31
Formel vægt 219.1904
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.