missing translation for 'onlineSavingsMsg'
Få mere at vide

Minocycline Hydrochloride Dihydrate, LoGiCal

Artikelnummer. 30462994
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30462994 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30462994 Leverandør LoGiCal Leverandørnr. LGCFOR1410.00

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Minocycline Hydrochloride Dihydrate
CAS 128420-71-3
Analyt- eller komponentnavne Minocycline Hydrochloride Dihydrate
Forsendelsestilstand Room Temperature
Molekylær formel C23 H27 N3 O7 . Cl H . 2 H2 O
Certificeringer/compliance ISO 9001
Synonym Minocycline Hydrochloride Dihydrate, (4S,4aS,5aR,12aS)-4,7-Bis(dimethylamino)-3,10,12,12a-tetrahydroxy-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide hydrochloride dihydrate, 2-Naphthacenecarboxamide, 4,7-bis(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,10,12,12a-tetrahydroxy-1,11-dioxo-, hydrochloride, hydrate (1:1:2), (4S,4aS,5aR,12aS)-, 2-Naphthacenecarboxamide, 4,7-bis(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,10,12,12a-tetrahydroxy-1,11-dioxo-, monohydrochloride, dihydrate, (4S,4aS,5aR,12aS)-, 2-Naphthacenecarboxamide, 4,7-bis(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,10,12,12a-tetrahydroxy-1,11-dioxo-, monohydrochloride, dihydrate, [4S-(4alpha,4aalpha,5aalpha,12aalpha)]-, Tigecycline Imp. C (EP) as Hydrochloride Dihydrate, Tigecycline Impurity C as Hydrochloride Dihydrate
InChI formel InChI=1S/C23H27N3O7.ClH.2H2O/c1-25(2)12-5-6-13(27)15-10(12)7-9-8-11-17(26(3)4)19(29)16(22(24)32)21(31)23(11,33)20(30)14(9)18(15)28;;;/h5-6,9,11,17,27,29-30,33H,7-8H2,1-4H3,(H2,24,32);1H;2*1H2/t9-,11-,17-,23-;;;/m0.../s1
Løsningstype Neat
SMIL O.O.Cl.CN(C)[C@H]1[C@@H]2C[C@@H]3Cc4c(ccc(O)c4C(=O)C3=C(O)[C@]2(O)C(=O)C(=C1O)C(=O)N)N(C)C
IUPAC navn (4S,4aS,5aR,12aS)-4,7-bis(dimethylamino)-3,10,12,12a-tetrahydroxy-1,11-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;dihydrate;hydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 529.97
Mængde 10 mg
Formel vægt 529.1827 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.