Learn More
Brady™ MiniMark™ Ink Ribbons
These ribbons have standard scratch, smear and chemical resistance. MiniMark™ Ink Ribbons for B-7951 and B-7589 materials.
Brand: Brady™ 710195
Additional Details : Weight : 0.12000kg
Description
- These ribbons have standard scratch, smear and chemical resistance.
- MiniMark ribbon for B-7951 and B-7589 materials.
Specifications
Printer Ribbon | |
MiniMark Printer | |
4.33 in. | |
110 mm | |
90 m x 110 mm | |
Ink side out |
Blue | |
98.4 yd. | |
90 m | |
12.7 mm I.D. | |
110 mm Blue Ribbon (1 Ea.) | |
1 Ea. |