missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kartell™ MiniFix Micropipettor
An innovative product line of fixed-volume air dispacement mini-micropipettes. Kartell™ MiniFix Micropipettor is designed for fast, repetitive, and precise operation and are competitively priced.
Brand: Kartell™ 0128500
Additional Details : Weight : 0.40000kg
Description
- Fully autoclavable
- Designed for fast, repetitive and precise operation
- Very compact in terms of size and weight
- Designed for routine or biological hazard work
- Kartell™ tips fit on the conical shaft as well as most other commercially available tips
- The plunger button has a different color based on dosing capacity
- Individually calibrated in compliance with ISO8655, the new international standard for piston-operated volumetric apparatus
- Carries the CE conformity mark
Specifications
±1.5% | |
Kartell™ 956/959 | |
Red | |
1 | |
5 μL | |
1/Pk. | |
10 g |
CE/IVD | |
Autoclavable | |
in Situ Analyses and Sampling, Routine or Biological Hazard Work | |
Mini Micropipettor | |
Routine or biological hazard work, in situ analyses and sampling | |
13 cm |