Learn More
FBPase 1 Antibody, PerCP, R&D Systems™
Product Code. 18604632 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.1mL
Product Code. Unit Size
18604632 0.1mL
1 options
Product Code. 18604632 Supplier Novus Biologicals Supplier No. NBP297978PCP
 Restricted Item
Forventet forsendelse: 10-12-2026
A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody
FBPase 1 Polyclonal specifically detects FBPase 1 in Human samples. It is validated for Immunohistochemistry/Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY
Specifications
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Product Type Antibody
Product Line Novus